Assay Details
Gene Symbol: RAB11FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192771.1 | NP_001179700.1 | 2 - 3 | 1093 | 76 | |
GenBank mRNA | BC151810.1 | - | 2 - 3 | 1093 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
