Assay Details
Gene Symbol: NES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206591.1 | NP_001193520.1 | 3 - 4 | 977 | 70 | |
GenBank mRNA | AB257750.1 | - | 2 - 3 | 159 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
