Assay Details
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206597.1 | NP_001193526.1 | 3 - 4 | 461 | 65 | |
GenBank mRNA | DV916511.1 | - | 1 - 2 | 151 | 65 | |
DV917166.1 | - | 4 - 5 | 443 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
