Assay Details
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206610.1 | NP_001193539.1 | 1 - 2 | 176 | 94 | |
GenBank mRNA | BT025443.1 | - | 2 - 3 | 193 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
