Assay Details
Gene Symbol: STIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206662.2 | NP_001193591.1 | 14 - 15 | 2826 | 97 | |
GenBank mRNA | CK776513.1 | - | 1 - 2 | 112 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
