Assay Details
Gene Symbol: FGF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206678.1 | NP_001193607.1 | 2 - 3 | 266 | 97 | |
GenBank mRNA | CN433133.1 | - | 2 - 3 | 270 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
