Assay Details
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206698.2 | NP_001193627.1 | 3 - 4 | 843 | 105 | |
GenBank mRNA | BC114070.1 | - | 3 - 4 | 844 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
