Assay Details
Gene Symbol: PKDCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206721.1 | NP_001193650.1 | 4 - 5 | 1276 | 98 | |
GenBank mRNA | EH181331.1 | - | 3 - 4 | 376 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
