Assay Details
Gene Symbol: ENPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206724.1 | NP_001193653.1 | 2 - 3 | 890 | 64 | |
GenBank mRNA | DV913973.1 | - | 1 - 2 | 438 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
