Assay Details
Gene Symbol: IGSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206778.1 | NP_001193707.1 | 2 - 3 | 244 | 118 | |
NM_001206779.1 | NP_001193708.1 | 2 - 3 | 244 | 118 | ||
NM_001206780.1 | NP_001193709.1 | 2 - 3 | 244 | 118 | ||
GenBank mRNA | CK949676.1 | - | 2 - 3 | 226 | 118 | |
CK951630.1 | - | 2 - 3 | 252 | 118 | ||
EV635438.1 | - | 2 - 3 | 208 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206778.1, NM_001206779.1, NM_001206780.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
