Assay Details
Gene Symbol: IGSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206785.1 | NP_001193714.1 | 4 - 5 | 5026 | 85 | |
GenBank mRNA | EE358837.1 | - | 1 - 2 | 526 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
