Assay Details
Gene Symbol: TCTEX1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166555.1 | NP_001160027.1 | 2 - 3 | 398 | 98 | |
GenBank mRNA | BC108235.1 | - | 2 - 3 | 384 | 98 | |
DV832959.1 | - | 2 - 3 | 376 | 98 | ||
DV846623.1 | - | 2 - 3 | 399 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
