Assay Details
Gene Symbol: TCTEX1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166555.1 | NP_001160027.1 | 3 - 4 | 461 | 79 | |
GenBank mRNA | BC108235.1 | - | 3 - 4 | 447 | 79 | |
DV832959.1 | - | 3 - 4 | 439 | 79 | ||
DV846623.1 | - | 3 - 4 | 462 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
