Assay Details
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167896.2 | NP_001161368.1 | 7 - 8 | 1333 | 93 | |
GenBank mRNA | BC105241.1 | - | 7 - 8 | 1024 | 93 | |
BC150062.1 | - | 7 - 8 | 1332 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
