Assay Details
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191208.1 | NP_001178137.1 | 1 - 2 | 72 | 78 | |
GenBank mRNA | DY134218.1 | - | 1 - 2 | 79 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191208.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
