Assay Details
Gene Symbol: ELK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191236.1 | NP_001178165.1 | 1 - 2 | 83 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191236.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
