Assay Details
Gene Symbol: HRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191241.2 | NP_001178170.1 | 1 - 2 | 461 | 103 | |
GenBank mRNA | EV627889.1 | - | 1 - 2 | 464 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
