Assay Details
Gene Symbol: NIBAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191282.1 | NP_001178211.1 | 10 - 11 | 1460 | 71 | |
GenBank mRNA | BC148868.1 | - | 10 - 11 | 1433 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
