Assay Details
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206815.1 | NP_001193744.1 | 1 - 2 | 136 | 86 | |
GenBank mRNA | EV631432.1 | - | 1 - 2 | 139 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206815.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
