Assay Details
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206815.1 | NP_001193744.1 | 4 - 5 | 1283 | 105 | |
GenBank mRNA | EV704037.1 | - | 2 - 3 | 174 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
