Assay Details
Gene Symbol: LOC508666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244199.1 | NP_001231128.1 | 1 - 2 | 396 | 109 | |
GenBank mRNA | BC110158.1 | - | 1 - 2 | 366 | 109 | |
EV633346.1 | - | 1 - 2 | 399 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244199.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
