Assay Details
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191339.2 | NP_001178268.2 | 4 - 5 | 943 | 83 | |
GenBank mRNA | AW658474.1 | - | 1 - 2 | 195 | 83 | |
BC149915.1 | - | 2 - 3 | 456 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
