Assay Details
Gene Symbol: NSUN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191381.1 | NP_001178310.1 | 4 - 5 | 781 | 87 | |
GenBank mRNA | BT026151.1 | - | 4 - 5 | 730 | 87 | |
DV819148.1 | - | 4 - 5 | 733 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
