Assay Details
Gene Symbol: GPR137B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191481.1 | NP_001178410.1 | 1 - 2 | 502 | 89 | |
GenBank mRNA | DV806566.1 | - | 1 - 2 | 486 | 89 | |
EH127404.1 | - | 1 - 2 | 503 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
