Assay Details
Gene Symbol: GPR137B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191481.1 | NP_001178410.1 | 2 - 3 | 552 | 101 | |
GenBank mRNA | DV806566.1 | - | 2 - 3 | 536 | 101 | |
EB726511.1 | - | 1 - 2 | 60 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
