Assay Details
Gene Symbol: GSDMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244217.1 | NP_001231146.1 | 2 - 3 | 250 | 64 | |
GenBank mRNA | BC149305.1 | - | 2 - 3 | 251 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244217.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
