Assay Details
Gene Symbol: PGAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192191.1 | NP_001179120.1 | 3 - 4 | 542 | 75 | |
GenBank mRNA | BC149319.1 | - | 3 - 4 | 534 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
