Assay Details
Gene Symbol: P2RY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192295.1 | NP_001179224.1 | 3 - 4 | 271 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001192295.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
