Assay Details
Gene Symbol: MXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192380.2 | NP_001179309.1 | 4 - 5 | 314 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
