Assay Details
Gene Symbol: MRPL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192391.1 | NP_001179320.1 | 1 - 2 | 62 | 85 | |
GenBank mRNA | BC103008.1 | - | 1 - 2 | 41 | 85 | |
BT025388.1 | - | 1 - 2 | 40 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
