Assay Details
Gene Symbol: MRPL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192391.1 | NP_001179320.1 | 3 - 4 | 393 | 100 | |
GenBank mRNA | BC103008.1 | - | 3 - 4 | 372 | 100 | |
BT025388.1 | - | 3 - 4 | 371 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
