Assay Details
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192981.2 | NP_001179910.1 | 4 - 5 | 551 | 61 | |
GenBank mRNA | BC126535.1 | - | 4 - 5 | 561 | 61 | |
BC151655.1 | - | 4 - 5 | 561 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
