Assay Details
Gene Symbol: PLA2G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193052.1 | NP_001179981.1 | 2 - 3 | 309 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193052.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
