Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193109.2 | NP_001180038.2 | 3 - 4 | 454 | 84 | |
GenBank mRNA | EV626017.1 | - | 3 - 4 | 416 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
