Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193109.2 | NP_001180038.2 | 9 - 10 | 1222 | 91 | |
GenBank mRNA | DV888705.1 | - | 1 - 2 | 125 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
