Assay Details
Gene Symbol: DND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193145.1 | NP_001180074.1 | 2 - 3 | 143 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
