Assay Details
Gene Symbol: ARPC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014844.3 | NP_001014844.1 | 8 - 9 | 1043 | 68 | |
GenBank mRNA | BC102942.1 | - | 8 - 9 | 1044 | 68 | |
BT021526.1 | - | 7 - 8 | 996 | 68 | ||
BT021895.1 | - | 7 - 8 | 990 | 68 | ||
BT025344.1 | - | 7 - 8 | 984 | 68 | ||
BT026160.1 | - | 7 - 8 | 985 | 68 | ||
BT026274.1 | - | 7 - 8 | 984 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
