Assay Details
Gene Symbol: ASMTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035058.1 | NP_001030230.1 | 7 - 8 | 927 | 61 | |
GenBank mRNA | BC102999.1 | - | 7 - 8 | 927 | 61 | |
BT026261.1 | - | 7 - 8 | 951 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
