Assay Details
Gene Symbol: NPY1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045898.1 | NP_001039363.1 | 1 - 2 | 179 | 129 | |
GenBank mRNA | BC114695.1 | - | 1 - 2 | 179 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114695.1, NM_001045898.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
