Assay Details
Gene Symbol: RASSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076105.1 | NP_001069573.1 | 4 - 5 | 1002 | 115 | |
GenBank mRNA | BC116069.1 | - | 4 - 5 | 1002 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
