Assay Details
Gene Symbol: RAB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076233.1 | NP_001069701.1 | 2 - 3 | 137 | 77 | |
GenBank mRNA | BC105393.1 | - | 2 - 3 | 137 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
