Assay Details
Gene Symbol: TRAPPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076413.2 | NP_001069881.1 | 2 - 3 | 267 | 100 | |
GenBank mRNA | BC118345.1 | - | 2 - 3 | 268 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
