Assay Details
Gene Symbol: PRKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199065.1 | NP_001185994.1 | 9 - 10 | 1143 | 61 | |
GenBank mRNA | FJ555558.1 | - | 9 - 10 | 1078 | 61 | |
FJ555559.1 | - | 8 - 9 | 962 | 61 | ||
FJ555560.1 | - | 7 - 8 | 853 | 61 | ||
GU290223.1 | - | 7 - 8 | 805 | 61 | ||
GU290224.1 | - | 7 - 8 | 832 | 61 | ||
GU290226.1 | - | 8 - 9 | 1041 | 61 | ||
GU322361.1 | - | 9 - 10 | 1132 | 61 | ||
GU322362.1 | - | 8 - 9 | 969 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
