Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205379.1 | NP_001192308.1 | 6 - 7 | 799 | 66 | |
GenBank mRNA | BC103103.1 | - | 6 - 7 | 781 | 66 | |
DT808282.1 | - | 3 - 4 | 542 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
