Assay Details
Gene Symbol: MIR221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030881.1 | 1 | 47 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-222 | MIR222 |
MIRN222, bta-mir-222, mir-222 |
-585.0 | Sense | Chr. X - 98125921 - 98126030 on NCBI Build ARS-UCD1.2 |
Downstream | divergent protein kinase domain 2B | DIPK2B |
CXHXorf36, CXORF36 |
430.0kb | Sense | Chr. X - 98556745 - 98608340 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
