Assay Details
Gene Symbol: MIR222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030882.1 | 1 | 48 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 674 | ZNF674 | -657.4kb | Anti-sense | Chr. X - 97434264 - 97468519 on NCBI Build ARS-UCD1.2 | |
Downstream | microRNA mir-221 | MIR221 |
MIRN221, bta-mir-221, mir-221 |
585.0 | Sense | Chr. X - 98126615 - 98126724 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
