Assay Details
Gene Symbol: MIR2355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031074.2 | 1 | 42 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carboxypeptidase O | CPO | -133.3kb | Anti-sense | Chr. 2 - 95234725 - 95266753 on NCBI Build ARS-UCD1.2 | |
Downstream | cAMP responsive element binding protein 1 | CREB1 | CREB | 464.6kb | Anti-sense | Chr. 2 - 95864818 - 95897170 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
