Assay Details
Gene Symbol: MIR2284U | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031075.1 | 1 | 43 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | complement C1q A chain | C1QA | -108.2kb | Anti-sense | Chr. 2 - 130189865 - 130192754 on NCBI Build ARS-UCD1.2 | |
Downstream | cell division cycle 42 | CDC42 | 432.6kb | Anti-sense | Chr. 2 - 130733638 - 130754601 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
