Assay Details
Gene Symbol: MIR654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031138.1 | 1 | 35 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-376d | MIR376D | bta-mir-376d | -72.0 | Sense | Chr. 21 - 65929322 - 65929403 on NCBI Build ARS-UCD1.2 |
Downstream | microRNA mir-376b | MIR376B | bta-mir-376b | 143.0 | Sense | Chr. 21 - 65929698 - 65929790 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
