Assay Details
Gene Symbol: MIR28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031148.1 | 1 | 47 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tumor protein p63 regulated 1 | TPRG1 | -492.2kb | Sense | Chr. 1 - 78014339 - 78169293 on NCBI Build ARS-UCD1.2 | |
Downstream | BCL6 transcription repressor | BCL6 | 897.1kb | Anti-sense | Chr. 1 - 79558619 - 79611185 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
